Learn More
RPS25 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 155661AP
Catalog No. 50-173-0319
Add to cart
Description
Description
15566-1-AP is a rabbit polyclonal antibody raised against full-length human RPS25.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S25E family of ribosomal proteins. It is located in the cytoplasm.
Specifications
Specifications
RPS25
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P62851
RPS25
RPS25 Fusion Protein Ag7825
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Immunoprecipitation, Western Blot
Unconjugated
RPS25
Liquid
Rabbit
IgG
150 μL
-20°C
Primary
6230