Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPS26 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313001
This item is not returnable.
View return policy
Description
RPS26 Polyclonal antibody specifically detects RPS26 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
RPS26 | |
Polyclonal | |
PBS, 20% Glycerol (pH7) | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
6231 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
40S ribosomal protein S26, DBA10, MGC104292, ribosomal protein S26 | |
Recombinant protein encompassing a sequence within the N-terminus region of human RPS26. The exact sequence is proprietary. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction