Learn More
RPS27 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 153551AP
Catalog No. 50-173-0172
Add to cart
Description
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S27E family of ribosomal proteins. It contains a C4-type zinc finger domain that can bind to zinc. The encoded protein has been shown to be able to bind to nucleic acid. It is located in the cytoplasm as a ribosomal component, but it has also been detected in the nucleus. Studies in rat indicate that ribosomal protein S27 is located near ribosomal protein S18 in the 40S subunit and is covalently linked to translation initiation factor eIF3. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
Specifications
RPS27
0.49 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P42677, Q6ZWU9, Q71TY3
40S ribosomal protein S27, Metallopan stimulin 1, MPS 1, MPS1, ribosomal protein S27, RPS27
Rabbit
IgG
150 μL
-20°C
Primary
57294, 6232, 94266
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RPS27
Liquid
RPS27
RPS27 Fusion Protein Ag7601
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat