Learn More
RPS27A Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 149461AP
Catalog No. 50-172-9855
Add to cart
Description
Description
Ubiquitin, a highly conserved protein that has a major role in targeting cellular proteins for degradation by the 26S proteosome, is synthesized as a precursor protein consisting of either polyubiquitin chains or a single ubiquitin fused to an unrelated protein. This gene encodes a fusion protein consisting of ubiquitin at the N terminus and ribosomal protein S27a at the C terminus. When expressed in yeast, the protein is post-translationally processed, generating free ubiquitin monomer and ribosomal protein S27a. Ribosomal protein S27a is a component of the 40S subunit of the ribosome and belongs to the S27AE family of ribosomal proteins. It contains C4-type zinc finger domains and is located in the cytoplasm. Pseudogenes derived from this gene are present in the genome. As with ribosomal protein S27a, ribosomal protein L40 is also synthesized as a fusion protein with ubiquitin; similarly, ribosomal protein S30 is synthesized as a fusion protein with the ubiquitin-like protein fubi. Multiple alternatively spliced transcript variants that encode the same proteins have been identified.
Specifications
Specifications
RPS27A
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P62979, P62983
CEP80, HUBCEP80, ribosomal protein S27a, RPS27A, UBA80, UBCEP1, UBCEP80, ubiquitin
Rabbit
IgG
150 μL
-20°C
Primary
6233, 78294
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RPS27A
Liquid
RPS27A
RPS27A Fusion Protein Ag6750
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse