Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPS4X Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549949
Description
The antibody detects endogenous levels of total RPS4X protein.
RPS4X Polyclonal antibody specifically detects RPS4X in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
RPS4X | |
Polyclonal | |
Unconjugated | |
RPS4X | |
40S ribosomal protein S4, X isoform; CCG2; cell cycle gene 2; DXS306; LOC100362640; ribosomal protein S4, X-linked; ribosomal protein S4, X-linked X-like; ribosomal protein S4X isoform; RPS4; Rps4-1; Rps4x; S4; SCAR; SCR10; single copy abundant mRNA protein; single-copy abundant mRNA; Small ribosomal subunit protein eS4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100362640, 20102, 6191 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P62701, P62702, P62703 | |
RPS4X | |
Synthesized peptide derived from internal of human RPS4X. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction