Learn More
RPS6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 148231AP
Catalog No. 50-172-9753
Add to cart
Description
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a cytoplasmic ribosomal protein that is a component of the 40S subunit. The protein belongs to the S6E family of ribosomal proteins. It is the major substrate of protein kinases in the ribosome, with subsets of five C-terminal serine residues phosphorylated by different protein kinases. Phosphorylation is induced by a wide range of stimuli, including growth factors, tumor-promoting agents, and mitogens. Dephosphorylation occurs at growth arrest. The protein may contribute to the control of cell growth and proliferation through the selective translation of particular classes of mRNA. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
Specifications
RPS6
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P62753, P62754, P62755
40S ribosomal protein S6, Phosphoprotein NP33, ribosomal protein S6, RPS6
Rabbit
IgG
150 μL
-20°C
Primary
20104, 29304, 6194
Immunoprecipitation, Western Blot
Unconjugated
RPS6
Liquid
RPS6
RPS6 Fusion Protein Ag6599
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat