Learn More
RPS6KA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 164631AP
Catalog No. 50-173-0961
Add to cart
Description
Description
This gene encodes a zinc finger protein of unknown function. It bears similarity to a zinc finger protein which acts as a transcriptional activator. This gene lies in a region of the X chromosome which has been associated with mental retardation.
Specifications
Specifications
RPS6KA1
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P18653, Q15418, Q63531
HU 1, MAPKAP kinase 1a, MAPKAPK 1a, MAPKAPK1A, p90 RSK 1, p90RSK1, p90S6K, Ribosomal S6 kinase 1, RPS6KA1, RSK, RSK 1, RSK1, S6K alpha 1
Rabbit
IgG
150 μL
-20°C
Primary
20111, 6195, 81771
Western Blot
Unconjugated
RPS6KA1
Liquid
RPS6KA1
RPS6KA1 Fusion Protein Ag9537
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat