Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Human RPS6KA3 (RSK2), His Tag Recombinant Protein
Human RPS6KA3 (RSK2), His Tag Recombinant Protein
$562.00
Specifications
Accession Number | NP_004577 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 86.3 kDa |
Gene Alias | RPS6KA3 (RSK2) |
KinaseFamily | RSK (RPS6K) Kinase Family |
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
PV3323
|
Thermo Scientific™
PV3323 |
10μg | 10μg |
Each of 1 for $562.00
|
|
||||
Description
RPS6KA3 (RSK2) is a serine⁄threonine kinase that modulates many cellular processes including transcription, adhesion, growth, survival, and proliferation.Specifications
NP_004577 | |
86.3 kDa | |
RSK (RPS6K) Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris pH 7.5, 150mM NaCl, 0.05mM EDTA, 0.05% Triton X-10, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Baculovirus |
Kinase Assay | |
RPS6KA3 (RSK2) | |
Human | |
His Tag | |
Dry Ice |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title