Learn More
Abnova™ RPS6KA5 (Human) Recombinant Protein
For use with Functional Study.
Manufacturer:  Abnova™ P6524
Catalog No. 89-339-466
Add to cart
Description
Description
Human RPS6KA5 (NP_004746.2, 1 a.a. - 802 a.a.) full length recombinant protein with GST-tag at N-terminal using baculovirus expression system.
Specifications
Specifications
Functional Study
ribosomal protein S6 kinase, 90kDa, polypeptide 5
5 µg
MGC1911, MSK1, MSPK1, RLPK
9252
The purity was assessed by SDS-PAGE/CBB staining.
Stored at -80°C. Aliquot to avoid repeated freezing and thawing.