Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPS6KB2 Mouse anti-Human, Mouse, Clone: 164CT21.2.2, Abnova™

Mouse monoclonal antibody raised against human RPS6KB2.

Manufacturer:  Abnova Corporation MAB12306

Catalog No. 89-934-874


Add to cart

Description

Description

This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates the S6 ribosomal protein and eucaryotic translation initiation factor 4B (eIF4B). Phosphorylation of S6 leads to an increase in protein synthesis and cell proliferation. [provided by RefSeq]

Specifications

Specifications

RPS6KB2
164CT21.2.2
Mouse monoclonal antibody raised against human RPS6KB2.
In ascites (0.09% sodium azide)
KLS/P70-beta/P70-beta-1/P70-beta-2/S6K-beta2/S6K2/SRK/STK14B/p70(S6K)-beta/p70S6Kb
Mouse
IgG1, kappa
RUO
Primary
6199
Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (1:10-1:50) Immunohistochemistry (1:50-1:100) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user.
Liquid
RPS6KB2
Recombinant His fusion protein corresponding to human RPS6KB2.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Murine
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit