Learn More
RPS6KC1 Mouse anti-Human, Clone: OTI10F11, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808825
Catalog No. 50-166-9106
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
May be involved in transmitting sphingosine-1 phosphate (SPP)-mediated signaling into the cell.
Specifications
Specifications
RPS6KC1
Monoclonal
Unconjugated
RPS6KC1
Lyophilized
RPS6KC1
Human recombit protein fragment corresponding to amino acids 1-305 of human RPS6KC1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI10F11
PBS with 8% trehalose and no preservative; pH 7.3
Q96S38
RPK118, RSKL1, S6K-delta-1, S6PKh1, humS6PKh1
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
26750