Learn More
Invitrogen™ RPS6KC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589452
Description
Immunogen sequence: DSSALGAVAS DSEQSKTEEE RESRSLFPGS LKPKLGKRDY LEKAGELIKL ALKKEEEDDY EAASDFYRKG VDLLLEGVQG ESSPTRREAV KRRTAEYLMR AESISSLYGK PQLDDVSQPP GSLSSRPLWN LRSPAEELKA FRVLGVIDKV LLVMDTRTEQ TFILKGLRKS SEYSRNRKTI IPRCVPNMVC LHKYIISEES V; Positive Samples: HeLa, LO2, 293T; Cellular Location: Cytoplasm, Membrane.
May be involved in transmitting sphingosine-1 phosphate (SPP)-mediated signaling into the cell. Plays a role in the recruitment of PRDX3 to early endosomes.
Specifications
RPS6KC1 | |
Polyclonal | |
Unconjugated | |
RPS6KC1 | |
52 kDa ribosomal protein S6 kinase; AA682037; B130003F20Rik; C80612; humS6PKh1; ribosomal protein S6 kinase C1; ribosomal protein S6 kinase delta-1; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase, 52kDa, polypeptide 1; ribosomal S6 kinase-like protein with two PSK domains 118 kDa protein; RPK118; Rps6kc1; RSKL1; S6K-delta-1; S6PKh1; SPHK1-binding protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
26750 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.63 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q96S38 | |
RPS6KC1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 200-400 of human RPS6KC1 (NP_0365562). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.