Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPS6KL1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548172
Description
Specifications
RPS6KL1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9Y6S9 | |
RPS6KL1 | |
This RPS6KL1 antibody is generated from rabbits immunized with a recombit protein of human RPS6KL1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RPS6KL1 | |
Ribosomal protein S6 kinase-like 1, RPS6KL1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83694 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction