Learn More
RPS9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA549950
Catalog No. PA549950
Add to cart
Description
Description
The antibody detects endogenous levels of total RPS9 protein.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S4P family of ribosomal proteins. It is located in the cytoplasm. Variable expression of this gene in colorectal cancers compared to adjacent normal tissues has been observed, although no correlation between the level of expression and the severity of the disease has been found. As is typical for genes encoding ribosomal proteins, multiple processed pseudogenes derived from this gene are dispersed through the genome.
Specifications
Specifications
RPS9
Polyclonal
Unconjugated
RPS9
Liquid
RPS9
Synthesized peptide derived from internal of human RPS9.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P46781, Q6ZWN5, P29314
40S ribosomal protein S9, ribosomal protein S9, RS9,
Rabbit
IgG
100 μL
-20°C
Primary
6203, 76846, 81772