Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPUSD4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113170
Description
RPUSD4 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Specifications
RPUSD4 | |
Polyclonal | |
Unconjugated | |
RPUSD4 | |
2410001E19Rik; Mitochondrial RNA pseudouridine synthase Rpusd4; RGD1311268; RNA pseudouridylate synthase domain containing 4; RNA pseudouridylate synthase domain-containing protein 4; Rpusd4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84881 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q96CM3 | |
RPUSD4 | |
Recombinant Human Mitochondrial RNA pseudouridine synthase RPUSD4 protein (41-240AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction