Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RRAD Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537885
Description
This antibody is predicted to react with human, rat, dog, pig and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
The Ras-encoded family of proteins bind GDP and GTP with high affinity. They possess a low level of intrinsic GTPase activity that increases more than 100-fold when interacting with cytosolic GTPase activating protein (GAP). Ras family members include H-Ras, K-Ras, N-Ras, M-Ras, R-Ras, ERas, Rheb, TC 21, RASL11B and Rad (Ras associated with diabetes) GTPase.Rad GTPase is a GTP-binding protein that is similar to Ras but has unique features. Unlike other small GTPases, Rad GTPase lacks typical pren-ylation motifs at its C terminus. The Rad GTPase enzyme binds calmodulin, inhibits vascular lesion formation, has low intrinsic GTPase activity and cannot be stimulated by any known GAP molecules. Rad GTPase is expressed in skeletal muscle, cardiac muscle and lung tissues and is overexpressed in the skeletal muscle tissue of individuals with type II diabetes.
Specifications
RRAD | |
Polyclonal | |
Unconjugated | |
RRAD | |
GTP-binding protein RAD; RAD; RAD1; RAS (RAD and GEM) like GTP binding 3; ras associated with diabetes; Ras-like GTP-binding protein rad; Ras-related associated with diabetes; REM3; RRAD; RRAD, Ras related glycolysis inhibitor and calcium channel regulator; RRAD, Ras related glycolysis inihibitor and calcium channel regulator | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
56437 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O88667 | |
RRAD | |
Peptide with sequence C-HRRSMPVDERDLQ, from the internal region of RRAD (aa36-48). | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction