Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RRAGA Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen RRAGA
Applications Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093366
View Documents
Novus Biologicals
NBP304667100UL
100 μg Each for $320.71
Add to cart
 
NB093367
View Documents
Novus Biologicals
NBP30466720UL
20 μg Each for $138.37
Add to cart
 
Description

Description

RRAGA Polyclonal antibody specifically detects RRAGA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

RRAGA
Unconjugated
Rabbit
Cancer, mTOR Pathway
PBS with 50% glycerol, pH7.3.
10670
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, Western Blot
Purified
RUO
Human, Mouse, Rat
Adenovirus E3 14.7 kDa-interacting protein 1, FIP1, FIP-1adenovirus E3-14.7K interacting protein 1, Rag A, RagA, RAGAras-related GTP-binding protein A, Ras-related GTP binding A
Recombinant fusion protein containing a sequence corresponding to amino acids 1-313 of human RRAGA (NP_006561.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit