Learn More
RRAGC/RagC Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304299AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
RRAGC is a monomeric guanine nucleotide-binding protein, or G protein. By binding GTP or GDP, small G proteins act as molecular switches in numerous cell processes and signaling pathways.Specifications
RRAGC/RagC | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q99K70, Q9HB90 | |
Rragc | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RRAGC | |
GTPase-interacting protein 2; Rag C protein; TIB929; ras-related GTP-binding protein C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54170, 64121 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.