Learn More
RRBP1 Mouse anti-Human, Alexa Fluor™ 532, Clone: 5610, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243600AF532
Catalog No. NP24360A532
Add to cart
Description
Description
RRBP1 Monoclonal antibody specifically detects RRBP1 in Human samples. It is validated for Western Blot,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
RRBP1
5610
Western Blot, Immunocytochemistry/Immunofluorescence
180 kDa ribosome receptor homolog, DKFZp586A1420, ES/130, ES/130-related protein, ES130, FLJ36146, hES, KIAA1398, MGC157720, MGC157721, ribosome binding protein 1 (dog 180kD homolog), ribosome binding protein 1 homolog 180kDa (dog), Ribosome receptor protein, ribosome-binding protein 1, RRp
Mouse
IgG2a
Store at 4°C in the dark.
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
RRBP1
Recombinant protein encompassing a sequence within the C-terminus region of human Ribosome binding protein 1. The exact sequence is proprietary.
0.1 mL
Primary
6238