Learn More
RRM1 Mouse anti-Human, Clone: OTI13E4, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF801315
Catalog No. 50-166-5551
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
RRM1 (Ribonucleotide reductase M1 polypeptide) is one of two non-identical subunits that constitute ribonucleoside-diphosphate reductase, an enzyme which catalyzes the biosynthesis of deoxyribonucleotides from the corresponding ribonucleotides. It provides the precursors necessary for DNA synthesis. RRM1 is involved in carcinogenesis, tumor progression.
Specifications
Specifications
RRM1
Monoclonal
Unconjugated
RRM1
Lyophilized
RRM1
Human recombit protein fragment corresponding to amino acids 541-792 of human RRM1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI13E4
PBS with 8% trehalose and no preservative; pH 7.3
P23921
R1, RIR1, RR1
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
6240