Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RRM1 Rabbit anti-Human, Clone: RRM1/4372R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen RRM1
Clone RRM1/4372R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122607
View Documents
Novus Biologicals
NBP30756710UG
100 μg
Each for $406.52
Add to cart
 
NB122608
View Documents
Novus Biologicals
NBP30756720UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

RRM1 Monoclonal antibody specifically detects RRM1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

RRM1
0.2 mg/ml
Immunohistochemistry (Paraffin), Peptide Array
Purified
RUO
Human
EC 1.17.4.1, R1, ribonucleoside-diphosphate reductase large subunit, Ribonucleoside-diphosphate reductase subunit M1, Ribonucleotide reductase large subunit, ribonucleotide reductase M1, ribonucleotide reductase M1 polypeptide, ribonucleotide reductase, large subunit, ribonucleotide reductase, R1 subunit, RIR1, RR1
Synthetic peptide corresponding to residues within aa700-800 of human RRM1 (exact sequence is proprietary)
Monoclonal
Protein A or G purified
RRM1/4372R
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Unconjugated
Rabbit
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
10 mM PBS with 0.05% BSA
6240
IgG Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit