Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RRM1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$482.50

Specifications

Antigen RRM1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP158187
SDP
View Documents
Novus Biologicals
NBP158187
100 μL
Each of 1 for $482.50
Add to Cart
 
Description

Description

RRM1 Polyclonal specifically detects RRM1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

RRM1
Polyclonal
Purified
RUO
P23921
6240
Synthetic peptides corresponding to RRM1(ribonucleotide reductase M1 polypeptide) The peptide sequence was selected from the N terminal of RRM1. Peptide sequence ATGSYIAGTNGNSNGLVPMLRVYNNTARYVDQGGNKRPGAFAIYLEPWHL.
Primary
Western Blot
Unconjugated
Rabbit
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
EC 1.17.4.1, R1, ribonucleoside-diphosphate reductase large subunit, Ribonucleoside-diphosphate reductase subunit M1, Ribonucleotide reductase large subunit, ribonucleotide reductase M1, ribonucleotide reductase M1 polypeptide, ribonucleotide reductase, large subunit, ribonucleotide reductase, R1 subunit, RIR1, RR1
RRM1
IgG
Protein A purified
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit