Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RRP15 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259131

Catalog No. NBP259131


Add to cart

Description

Description

RRP15 Polyclonal antibody specifically detects RRP15 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

RRP15
Unconjugated
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
CGI-115, KIAA0507Ribosomal RNA-processing protein 15, MGC22291, ribosomal RNA processing 15 homolog (S. cerevisiae), RRP15-like protein
Rabbit
IgG
100 ul
Store at 4C.
Polyclonal
Human
Western Blot, Immunoprecipitation
Western Blot, Immunoprecipitation
Affinity Purified
RRP15
Maps to a region between 75 and 125
Immunogen affinity purified
RUO
Primary
51018
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only