Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RRP4 Rabbit anti-Human, Polyclonal, Invitrogen
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen PA564991
Description
Immunogen sequence: WKVETNSRLDS VLLLSSMNLP GGELRRRSAE DELAMRGFLQ EGDLISAEVQ AVFSDGAVSL HTRSLKYGKL GQGVLVQVSP SLVKRQKTH Highest antigen sequence identity to the following orthologs - mouse 100%, rat 98%.
Component of the exosome 3 ft.->5 ft. exoribonuclease complex, a complex that degrades inherently unstable mRNAs containing AU-rich elements (AREs) within their 3 ft. untranslated regions. Required for the 3 ft.processing of the 7S pre-RNA to the mature 5.8S rRNA. Has a 3 ft.-5 ft. exonuclease activity.Specifications
RRP4 | |
Polyclonal | |
Unconjugated | |
EXOSC2 | |
Liquid | |
EXOSC2 | |
Recombit Human RRP4 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Immunocytochemistry, Immunofluorescence | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13868 | |
RRP4; Rrp4p; hRrp4p; p7; exosome complex component RRP4; exosome complex exonuclease RRP4; homolog of yeast RRP4 (ribosomal RNA processing 4); 3′ 5′ exoribonuclease (RRP4); homolog of yeast RRP4 (ribosomal RNA processing 4); 3′-5′-exoribonuclease; ribosomal RNA-processing protein 4 | |
Rabbit | |
IgG | |
100 μL | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Primary | |
23404 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only