Learn More
RRP7A Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 244901AP
Catalog No. 50-173-4385
Add to cart
Description
Description
In cooperation with other chaperones, Hsp70s stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage.
Specifications
Specifications
RRP7A
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9D1C9, Q9Y3A4
BK126B4.3, CGI 96, Gastric cancer antigen Zg14, RRP7A
Rabbit
IgG
150 μL
-20°C
Primary
27341, 74778
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RRP7A
Liquid
Rrp7a
RRP7A Fusion Protein Ag21636
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse