Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RRP9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541796
Description
Peptide sequence: IPRAKKGAEG KPPGHSSHVL CMAISSDGKY LASGDRSKLI LIWEAQSCQH Sequence homology: Cow: 79%; Dog: 79%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 79%; Pig: 93%; Rabbit: 93%; Rat: 86%.
The Ku autoantigen is a heterodimer of 70kDa (p70) and approximately 80kDa (p80) proteins. The p70/p80 dimer is important for function of a 460kDa DNA-dependent protein kinase that phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. Ku protein plays a role in cell signaling, proliferation, DNA repair, replication, transcriptional activation, and apoptosis.
Specifications
RRP9 | |
Polyclonal | |
Unconjugated | |
RRP9 | |
55kDa; D19435; D9Wsu10e; ribosomal RNA processing 9, small subunit (SSU) processome component, homolog; ribosomal RNA processing 9, small subunit (SSU) processome component, homolog (yeast); ribosomal RNA processing 9, U3 small nucleolar RNA binding protein; RNA, U3 small nucleolar interacting protein 2; RNU3IP2; Rrp9; RRP9 homolog; RRP9, small subunit (SSU) processome component, homolog; RRP9, small subunit (SSU) processome component, homolog (yeast); U3 small nucleolar ribonucleoprotein-associated 55 kDa protein; U3 small nucleolar RNA-interacting protein 2; U3 snoRNP-associated 55 kDa protein; U3 snoRNP-associated 55-kDa protein; U3 snoRNP-associated protein; U355K; U3-55k | |
Rabbit | |
Protein A | |
RUO | |
9136 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O43818 | |
RRP9 | |
Synthetic peptide directed towards the middle region of human RRP9. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction