Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RS 1

High affinity RAD51 activator; also CRISPR enhancer

$457.00

Specifications

Chemical Name or Material 4-Bromo-N-(4-bromophenyl)-3-[[(phenylmethyl)amino]sulfonyl]benzamide
Molecular Formula C20H16Br2N2O3S
CAS 312756-74-4
Product Type Target: DNA, RNA and Protein Synthesis Inhibitors
Formula Weight 524.23
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
581050 View Documents Tocris Bioscience™
5810/50
50mg Each for $457.00
Add to cart
 
581010 View Documents Tocris Bioscience™
5810/10
10mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

High affinity RAD51 activator (Kd range = 48-107 nM, value depends on cofactors present). Enhances homologous recombination activity of hRAD51. Enhances homology-directed repair ~2 to 5-fold and increases CRISPR-mediated knock-in efficiencies in vitro and in vivo.
Specifications

Specifications

4-Bromo-N-(4-bromophenyl)-3-[[(phenylmethyl)amino]sulfonyl]benzamide
312756-74-4
524.23
C20H16Br2N2O3S
Target: DNA, RNA and Protein Synthesis Inhibitors
Safety and Handling

Safety and Handling

Recommended Storage : Store at RT

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit