Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RS 17053 hydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

169505-93-5
C24H30Cl2N2O2
449.416
QFOPFGRPNPCPBX-UHFFFAOYSA-N
(N-[2-(2-Cyclopropylmethoxyphenoxy)ethyl]-5-chloro-α, α-dimethyl-1H-indole-3-ethanamine) hydrochloride
9824953

α1A antagonist

α1A-adrenoceptor antagonist, with very high affinity for α1A receptors (pKi and pA2 estimates of 9.1 - 9.9) and a 30 - 100 fold selectivity over the α1B and α1D subtypes (pKi and pA2 estimates 7.7 - 7.8).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
SMILES CC(C)(CC1=CNC2=C1C=C(C=C2)Cl)NCCOC3=CC=CC=C3OCC4CC4.Cl
IUPAC Name 1-(5-chloro-1H-indol-3-yl)-N-[2-[2-(cyclopropylmethoxy)phenoxy]ethyl]-2-methylpropan-2-amine;hydrochloride
Formula Weight 449.42
Chemical Name or Material RS 17053 hydrochloride
Grade >98%

Recommended Storage : Store at Room Temperature