Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RS 23597-190 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
A high affinity, selective competitive antagonist at 5-HT4 receptors.
Specifications
Quantity | 10 mg |
Synonym | 3-piperidin-1-yl propyl 4-amino-5-chloro-2-methoxybenzoate hydrochloride, rs 23597-190 hydrochloride, 3-piperidine-1-yl propyl 4-amino-5-chloro-2-methoxybenzoate hydrochloride, 3-piperidin-1-yl propyl4-amino-5-chloro-2-methoxybenzoate hydrochloride, 3-piperidine-1-yl propyl-4-amino-5-chloro-2-methoxybenzoate hydrochloride, 3-piperidin-1-ylpropyl 4-amino-5-chloro-2-methoxybenzoate hydrochloride, 3-piperidin-1-ylpropyl 4-azanyl-5-chloranyl-2-methoxy-benzoate hydrochloride, 4-amino-5-chloro-2-methoxy-benzoic acid 3-piperidinopropyl ester hydrochloride, 4-amino-5-chloro-2-methoxybenzoic acid 3-1-piperidinyl propyl ester hydrochloride, benzoic acid, 4-amino-5-chloro-2-methoxy-, 3-piperidine-1-yl propyl ester, hydrochloride |
Formula Weight | 363.28 |
Percent Purity | >98% |
Chemical Name or Material | RS 23597-190 hydrochloride |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction