RS 25344 hydrochloride, Tocris

Potent PDE4 inhibitor

Manufacturer: R&D Systems 3315/50

Quantity 50mg

 View more versions of this product

Catalog No. 33-155-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C19H13N5O4·HCl
    Formula Weight 411.8
    Synonym 1-(3-Nitrophenyl)-3-(4-pyridinylmethyl)-pyrido[2,3-d]pyrimidine-2,3-(1H,3H)-dione hydrochloride
    Chemical Name or Material RS 25344 hydrochloride

    Specifications

    Formula Weight 411.8
    Quantity 50mg
    Synonym 1-(3-Nitrophenyl)-3-(4-pyridinylmethyl)-pyrido[2,3-d]pyrimidine-2,3-(1H,3H)-dione hydrochloride
    Assay Percent Range >98%
    Molecular Formula C19H13N5O4·HCl
    CAS 152815-28-6
    Recommended Storage Desiccate at Room Temperature
    Chemical Name or Material RS 25344 hydrochloride

    Desiccate at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit