(RS)-3-Hydroxyphenylglycine, Tocris

Group I mGlu agonist

Manufacturer: R&D Systems 0324/50

Quantity 50mg

 View more versions of this product

Catalog No. 03-245-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C8H9NO3
    Formula Weight 167.16
    Chemical Name or Material (RS)-3-Hydroxyphenylglycine

    Specifications

    Quantity 50mg
    Formula Weight 167.16
    Assay Percent Range >98%
    Molecular Formula C8H9NO3
    CAS 31932-87-3
    Recommended Storage Desiccate at Room Temperature
    Chemical Name or Material (RS)-3-Hydroxyphenylglycine

    Desiccate at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit