Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(RS)-3-Hydroxyphenylglycine, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

31932-87-3
C8H9NO3
167.164
DQLYTFPAEVJTFM-UHFFFAOYSA-N
amino 3-hydroxyphenyl acetic acid,2-amino-2-3-hydroxyphenyl acetic acid,rs-3-hydroxyphenylglycine,chembl88124,rs-3h-pg,benzeneacetic acid, a-amino-3-hydroxy,3hpg,2-3-hydroxyphenyl glycine,amino 3-hydroxyphenyl acetic acid #,glycine, 2-m-hydroxyphenyl-, dl
1217
2-amino-2-(3-hydroxyphenyl)acetic acid
C1=CC(=CC(=C1)O)C(C(=O)O)N

Group I mGlu agonist

Agonist at PI-linked metabotropic glutamate receptors. Separate isomer (S)-3-Hydroxyphenylglycine (Cat. No. 0326) also available.
Formula Weight 167.16
Percent Purity >98%
Quantity 10mg
Chemical Name or Material (RS)-3-Hydroxyphenylglycine

Recommended Storage : Desiccate at Room Temperature