Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RS 39604 hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
167710-87-4
C26H37Cl2N3O6S
590.557
QSMYZGMJSGUWPM-UHFFFAOYSA-N
rs 39604 hydrochloride,gnf-pf-5475,n-2-4-3-4-amino-5-chloro-2-3,5-dimethoxybenzyl oxy phenyl-3-oxopropyl piperidin-1-yl ethyl methanesulfonamide hydrochloride,rs-39604 hydrochloride,1-4-amino-5-chloro-2-methoxyphenyl-3-1-2-methylsulphonylamino ethyl-4-piperidinyl-1-propanone hydrochloride,1-4-amino-5-chloro-2-3,5-dimethoxybenzyloxy phenyl-3-1-2-methylsulfonyl amino ethyl-4-piperidinyl-1-propanone hydrochloride,1-4-amino-5-chloro-2-3,5-dimethoxyphenyl methyloxy-3-1-2-methylsulphonylamino ethyl piperidin-4-yl propan-1-one hydrochloride
19081934
CHEBI:64081
N-[2-[4-[3-[4-amino-5-chloro-2-[(3,5-dimethoxyphenyl)methoxy]phenyl]-3-oxopropyl]piperidin-1-yl]ethyl]methanesulfonamide;hydrochloride
COC1=CC(=CC(=C1)COC2=CC(=C(C=C2C(=O)CCC3CCN(CC3)CCNS(=O)(=O)C)Cl)N)OC.Cl
167710-87-4
C26H37Cl2N3O6S
590.557
QSMYZGMJSGUWPM-UHFFFAOYSA-N
rs 39604 hydrochloride,gnf-pf-5475,n-2-4-3-4-amino-5-chloro-2-3,5-dimethoxybenzyl oxy phenyl-3-oxopropyl piperidin-1-yl ethyl methanesulfonamide hydrochloride,rs-39604 hydrochloride,1-4-amino-5-chloro-2-methoxyphenyl-3-1-2-methylsulphonylamino ethyl-4-piperidinyl-1-propanone hydrochloride,1-4-amino-5-chloro-2-3,5-dimethoxybenzyloxy phenyl-3-1-2-methylsulfonyl amino ethyl-4-piperidinyl-1-propanone hydrochloride,1-4-amino-5-chloro-2-3,5-dimethoxyphenyl methyloxy-3-1-2-methylsulphonylamino ethyl piperidin-4-yl propan-1-one hydrochloride
19081934
CHEBI:64081
N-[2-[4-[3-[4-amino-5-chloro-2-[(3,5-dimethoxyphenyl)methoxy]phenyl]-3-oxopropyl]piperidin-1-yl]ethyl]methanesulfonamide;hydrochloride
COC1=CC(=CC(=C1)COC2=CC(=C(C=C2C(=O)CCC3CCN(CC3)CCNS(=O)(=O)C)Cl)N)OC.Cl

5-HT4 antagonist

A potent and selective 5-HT4 antagonist, with a pKi of 9.1 at 5-HT4 receptors in guinea pig striatal membranes and greater than 1000-fold selectivity over 5-HT1A, 2C, 3 and D1, D2, M1, M2, AT1, B1 and α1C receptors. The ketone group gives RS 39604 a relatively long half life; it is also orally active and so suitable for in vivo studies.
Quantity 50mg
Formula Weight 590.56
Percent Purity >98%
Chemical Name or Material RS 39604 hydrochloride

Recommended Storage : Store at Room Temperature