Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RS 39604 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
A potent and selective 5-HT4 antagonist, with a pKi of 9.1 at 5-HT4 receptors in guinea pig striatal membranes and greater than 1000-fold selectivity over 5-HT1A, 2C, 3 and D1, D2, M1, M2, AT1, B1 and α1C receptors. The ketone group gives RS 39604 a relatively long half life; it is also orally active and so suitable for in vivo studies.
Specifications
Quantity | 10 mg |
Formula Weight | 590.56 |
Percent Purity | >98% |
Chemical Name or Material | RS 39604 hydrochloride |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction