Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RS 39604 hydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
167710-87-4
C26H37Cl2N3O6S
590.557
QSMYZGMJSGUWPM-UHFFFAOYSA-N
rs 39604 hydrochloride,gnf-pf-5475,n-2-4-3-4-amino-5-chloro-2-3,5-dimethoxybenzyl oxy phenyl-3-oxopropyl piperidin-1-yl ethyl methanesulfonamide hydrochloride,rs-39604 hydrochloride,1-4-amino-5-chloro-2-methoxyphenyl-3-1-2-methylsulphonylamino ethyl-4-piperidinyl-1-propanone hydrochloride,1-4-amino-5-chloro-2-3,5-dimethoxybenzyloxy phenyl-3-1-2-methylsulfonyl amino ethyl-4-piperidinyl-1-propanone hydrochloride,1-4-amino-5-chloro-2-3,5-dimethoxyphenyl methyloxy-3-1-2-methylsulphonylamino ethyl piperidin-4-yl propan-1-one hydrochloride
19081934
CHEBI:64081
N-[2-[4-[3-[4-amino-5-chloro-2-[(3,5-dimethoxyphenyl)methoxy]phenyl]-3-oxopropyl]piperidin-1-yl]ethyl]methanesulfonamide;hydrochloride
COC1=CC(=CC(=C1)COC2=CC(=C(C=C2C(=O)CCC3CCN(CC3)CCNS(=O)(=O)C)Cl)N)OC.Cl
167710-87-4
C26H37Cl2N3O6S
590.557
QSMYZGMJSGUWPM-UHFFFAOYSA-N
rs 39604 hydrochloride,gnf-pf-5475,n-2-4-3-4-amino-5-chloro-2-3,5-dimethoxybenzyl oxy phenyl-3-oxopropyl piperidin-1-yl ethyl methanesulfonamide hydrochloride,rs-39604 hydrochloride,1-4-amino-5-chloro-2-methoxyphenyl-3-1-2-methylsulphonylamino ethyl-4-piperidinyl-1-propanone hydrochloride,1-4-amino-5-chloro-2-3,5-dimethoxybenzyloxy phenyl-3-1-2-methylsulfonyl amino ethyl-4-piperidinyl-1-propanone hydrochloride,1-4-amino-5-chloro-2-3,5-dimethoxyphenyl methyloxy-3-1-2-methylsulphonylamino ethyl piperidin-4-yl propan-1-one hydrochloride
19081934
CHEBI:64081
N-[2-[4-[3-[4-amino-5-chloro-2-[(3,5-dimethoxyphenyl)methoxy]phenyl]-3-oxopropyl]piperidin-1-yl]ethyl]methanesulfonamide;hydrochloride
COC1=CC(=CC(=C1)COC2=CC(=C(C=C2C(=O)CCC3CCN(CC3)CCNS(=O)(=O)C)Cl)N)OC.Cl

5-HT4 antagonist

A potent and selective 5-HT4 antagonist, with a pKi of 9.1 at 5-HT4 receptors in guinea pig striatal membranes and greater than 1000-fold selectivity over 5-HT1A, 2C, 3 and D1, D2, M1, M2, AT1, B1 and α1C receptors. The ketone group gives RS 39604 a relatively long half life; it is also orally active and so suitable for in vivo studies.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 590.56
Percent Purity >98%
Chemical Name or Material RS 39604 hydrochloride

Recommended Storage : Store at Room Temperature