Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(RS)-4-Carboxy-3-hydroxyphenylglycine, Tocris Bioscience™

Catalog Number 031050
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
134052-66-7
C9H9NO5
211.173
GXZSAQLJWLCLOX-UHFFFAOYSA-N
4-chpgly, rs-4c3h-pg, 4-carboxy-3-hydroxyphenylglycine, 4-amino carboxy methyl-2-hydroxybenzoic acid, alpha-amino-4-carboxy-3-hydroxybenzeneacetic acid, +--alpha-amino-4-carboxy-3-hydroxybenzeneacetic acid, rs-4-carboxy-3-hydroxyphenylglycine, benzeneacetic acid, a-amino-4-carboxy-3-hydroxy, benzeneacetic acid, alpha-amino-4-carboxy-3-hydroxy, benzeneacetic acid, alpha-amino-4-carboxy-3-hydroxy-, +-
1297
4-[amino(carboxy)methyl]-2-hydroxybenzoic acid
C1=CC(=C(C=C1C(C(=O)O)N)O)C(=O)O
This item is not returnable. View return policy
134052-66-7
C9H9NO5
211.173
GXZSAQLJWLCLOX-UHFFFAOYSA-N
4-chpgly, rs-4c3h-pg, 4-carboxy-3-hydroxyphenylglycine, 4-amino carboxy methyl-2-hydroxybenzoic acid, alpha-amino-4-carboxy-3-hydroxybenzeneacetic acid, +--alpha-amino-4-carboxy-3-hydroxybenzeneacetic acid, rs-4-carboxy-3-hydroxyphenylglycine, benzeneacetic acid, a-amino-4-carboxy-3-hydroxy, benzeneacetic acid, alpha-amino-4-carboxy-3-hydroxy, benzeneacetic acid, alpha-amino-4-carboxy-3-hydroxy-, +-
1297
4-[amino(carboxy)methyl]-2-hydroxybenzoic acid
C1=CC(=C(C=C1C(C(=O)O)N)O)C(=O)O

Broad spectrum EAA ligand

Broad spectrum EAA agonist/antagonist.

S-enantiomer also available.

Formula Weight 211.17
Percent Purity >99%
Quantity 50 mg
Chemical Name or Material (RS)-4-Carboxy-3-hydroxyphenylglycine
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit