Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RS 45041-190 hydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

204274-74-8
C11H13Cl2N3
258.146
WRYJNVYEPVATQV-UHFFFAOYSA-N
4-Chloro-2-(imidazolin-2-yl)isoindoline hydrochloride
10332716

High affinity I2 ligand. Highly selective

Highly selective I2 imidazoline receptor ligand.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
SMILES C1CN=C(N1)N2CC3=C(C2)C(=CC=C3)Cl.Cl
IUPAC Name 4-chloro-2-(4,5-dihydro-1H-imidazol-2-yl)-1,3-dihydroisoindole;hydrochloride
Formula Weight 258.15
Chemical Name or Material RS 45041-190 hydrochloride
Grade >98%

Recommended Storage : Store at Room Temperature