Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RS 45041-190 hydrochloride, Tocris Bioscience™

Catalog Number 088950
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
204274-74-8
C11H13Cl2N3
258.146
WRYJNVYEPVATQV-UHFFFAOYSA-N
4-chloro-2-imidazolin-2-yl isoindoline hydrochloride, 4-chloro-2-4,5-dihydro-1h-imidazol-2-yl-1,3-dihydroisoindole hydrochloride, rs 45041-190 hydrochloride, 1h-isoindole,4-chloro-2-4,5-dihydro-1h-imidazol-2-yl-2,3-dihydro
10332716
4-chloro-2-(4,5-dihydro-1H-imidazol-2-yl)-1,3-dihydroisoindole;hydrochloride
C1CN=C(N1)N2CC3=C(C2)C(=CC=C3)Cl.Cl
This item is not returnable. View return policy
204274-74-8
C11H13Cl2N3
258.146
WRYJNVYEPVATQV-UHFFFAOYSA-N
4-chloro-2-imidazolin-2-yl isoindoline hydrochloride, 4-chloro-2-4,5-dihydro-1h-imidazol-2-yl-1,3-dihydroisoindole hydrochloride, rs 45041-190 hydrochloride, 1h-isoindole,4-chloro-2-4,5-dihydro-1h-imidazol-2-yl-2,3-dihydro
10332716
4-chloro-2-(4,5-dihydro-1H-imidazol-2-yl)-1,3-dihydroisoindole;hydrochloride
C1CN=C(N1)N2CC3=C(C2)C(=CC=C3)Cl.Cl

High affinity I2 ligand. Highly selective

Highly selective I2 imidazoline receptor ligand.
Quantity 50 mg
Formula Weight 258.15
Percent Purity >98%
Chemical Name or Material RS 45041-190 hydrochloride
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit