Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RS 56812 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
A highly potent and selective 5-HT3 partial agonist, with a pKi at 5-HT3 receptors in rat brain of 9.6, and greater than 1000-fold selectivity over a wide range of other neurotransmitter receptors.
Specifications
Quantity | 10 mg |
Formula Weight | 347.84 |
Percent Purity | >99% |
Chemical Name or Material | RS 56812 hydrochloride |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction