Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RS 67333 hydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
168986-60-5
C19H30Cl2N2O2
389.361
XVBGLZNYWUUAFF-UHFFFAOYSA-N
rs 67333 hydrochloride,1-4-amino-5-chloro-2-methoxyphenyl-3-1-butyl-4-piperidinyl-1-propanone hydrochloride,1-4-amino-5-chloro-2-methoxyphenyl-3-1-butylpiperidin-4-yl propan-1-one hydrochloride,rs-67333 hydrochloride,rs 67333 hydrochloride hplc,1-4-amino-5-chloro-2-methoxyphenyl-3-1-butyl-4-piperidinyl-1-propanone hcl
9800491
1-(4-amino-5-chloro-2-methoxyphenyl)-3-(1-butylpiperidin-4-yl)propan-1-one;hydrochloride
CCCCN1CCC(CC1)CCC(=O)C2=CC(=C(C=C2OC)N)Cl.Cl
168986-60-5
C19H30Cl2N2O2
389.361
XVBGLZNYWUUAFF-UHFFFAOYSA-N
rs 67333 hydrochloride,1-4-amino-5-chloro-2-methoxyphenyl-3-1-butyl-4-piperidinyl-1-propanone hydrochloride,1-4-amino-5-chloro-2-methoxyphenyl-3-1-butylpiperidin-4-yl propan-1-one hydrochloride,rs-67333 hydrochloride,rs 67333 hydrochloride hplc,1-4-amino-5-chloro-2-methoxyphenyl-3-1-butyl-4-piperidinyl-1-propanone hcl
9800491
1-(4-amino-5-chloro-2-methoxyphenyl)-3-(1-butylpiperidin-4-yl)propan-1-one;hydrochloride
CCCCN1CCC(CC1)CCC(=O)C2=CC(=C(C=C2OC)N)Cl.Cl

5-HT4 partial agonist

A potent and highly selective 5-HT4 partial agonist, with pKi values of 8.7 at 5-HT4 sites in guinea pig striatum and > 6 at various other receptors, including 5-HT1A, 1D, 2A, 2C, D1, D2 and M1-3. Active in vivo, with an intrinsic activity relative to 5-HT of 0.5.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 389.36
Percent Purity >99%
Chemical Name or Material RS 67333 hydrochloride

Recommended Storage : Store at Room Temperature