Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RS 67333 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
A potent and highly selective 5-HT4 partial agonist, with pKi values of 8.7 at 5-HT4 sites in guinea pig striatum and > 6 at various other receptors, including 5-HT1A, 1D, 2A, 2C, D1, D2 and M1-3. Active in vivo, with an intrinsic activity relative to 5-HT of 0.5.
Specifications
Quantity | 10 mg |
Formula Weight | 389.36 |
Percent Purity | >99% |
Chemical Name or Material | RS 67333 hydrochloride |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction