Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RS 67506 hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
168986-61-6
C18H29Cl2N3O4S
454.407
GFWKWEHKHLTRRF-UHFFFAOYSA-N
rs 67506 hydrochloride,1-4-amino-5-chloro-2-methoxyphenyl-3-1-2-methylsulphonylamino ethyl-4-piperidinyl-1-propanone hydrochloride,n-2-4-3-4-amino-5-chloro-2-methoxyphenyl-3-oxopropyl piperidin-1-yl ethyl methanesulfonamide hydrochloride,n-2-4-3-4-amino-5-chloro-2-methoxyphenyl-3-oxopropyl piperidin-1-yl ethyl methanesulfonamide hydrochloride,n-2-4-3-4-amino-5-chloro-2-methoxyphenyl-3-oxopropyl piperidin-1-yl ethyl methanesulfonamide hydrochloride,1-4-amino-5-chloro-2-methoxyphenyl-3-1-2-methyl sulphonylamino ethyl-4-piperidinyl-1-propanone hydrochloride,1-4-amino-5-chloro-2-methoxyphenyl-3-1-2-methylsulphonylamino ethyl-4-piperidinyl-1-propanone hydrochloride
133075
N-[2-[4-[3-(4-amino-5-chloro-2-methoxyphenyl)-3-oxopropyl]piperidin-1-yl]ethyl]methanesulfonamide;hydrochloride
COC1=CC(=C(C=C1C(=O)CCC2CCN(CC2)CCNS(=O)(=O)C)Cl)N.Cl
168986-61-6
C18H29Cl2N3O4S
454.407
GFWKWEHKHLTRRF-UHFFFAOYSA-N
rs 67506 hydrochloride,1-4-amino-5-chloro-2-methoxyphenyl-3-1-2-methylsulphonylamino ethyl-4-piperidinyl-1-propanone hydrochloride,n-2-4-3-4-amino-5-chloro-2-methoxyphenyl-3-oxopropyl piperidin-1-yl ethyl methanesulfonamide hydrochloride,n-2-4-3-4-amino-5-chloro-2-methoxyphenyl-3-oxopropyl piperidin-1-yl ethyl methanesulfonamide hydrochloride,n-2-4-3-4-amino-5-chloro-2-methoxyphenyl-3-oxopropyl piperidin-1-yl ethyl methanesulfonamide hydrochloride,1-4-amino-5-chloro-2-methoxyphenyl-3-1-2-methyl sulphonylamino ethyl-4-piperidinyl-1-propanone hydrochloride,1-4-amino-5-chloro-2-methoxyphenyl-3-1-2-methylsulphonylamino ethyl-4-piperidinyl-1-propanone hydrochloride
133075
N-[2-[4-[3-(4-amino-5-chloro-2-methoxyphenyl)-3-oxopropyl]piperidin-1-yl]ethyl]methanesulfonamide;hydrochloride
COC1=CC(=C(C=C1C(=O)CCC2CCN(CC2)CCNS(=O)(=O)C)Cl)N.Cl

5-HT4 partial agonist

A potent and selective 5-HT4 partial agonist, with a pKi of 8.8 at 5-HT4 sites in guinea pig striatum, but < 6.0 at several other receptors including 5-HT1A, 1D, 2A, 2C, D1, D2 and M1-3. It is active in vivo and has an intrinsic activity compared to 5-HT of 0.6.
Quantity 50mg
Formula Weight 454.41
Percent Purity >98%
Chemical Name or Material RS 67506 hydrochloride

Recommended Storage : Store at Room Temperature