Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RS 79948 hydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
186002-54-0
C19H29ClN2O3S
400.962
DZTZUOBWDBPPJQ-BQBHMPFISA-N
rs 79948 hydrochloride,8ar,12as,13as-5,8,8a,9,10,11,12,12a,13,13a-dechydro-3-methoxy-12-ethylsulfonyl-6h-isoquino 2,1-g 1,6 naphthyridine hydrochloride,4ar,12bs,13as-1-ethanesulfonyl-10-methoxy-2,3,4,4a,5,7,8,12b,13,13a-decahydro-1,6-diazatetraphene hydrochloride,8ar,12as,13as-12-ethylsulfonyl-5,8,8a,9,10,11,12,12a,13,13a-decahydro-3-methoxy-6h-isoquino 2,1-g 1,6 naphthyridine hydrochloride,8ar,12as,13as-5,8,8a,9,10,11,12,12a,13,13a-decahydro-3-methoxy-12-ethylsulfonyl-6h-isoquino 2,1-g 1,6 naphthyridine hydrochloride
9908991
(8aR,12aS,13aS)-12-ethylsulfonyl-3-methoxy-5,6,8,8a,9,10,11,12a,13,13a-decahydroisoquinolino[2,1-g][1,6]naphthyridine;hydrochloride
CCS(=O)(=O)N1CCCC2C1CC3C4=C(CCN3C2)C=C(C=C4)OC.Cl
186002-54-0
C19H29ClN2O3S
400.962
DZTZUOBWDBPPJQ-BQBHMPFISA-N
rs 79948 hydrochloride,8ar,12as,13as-5,8,8a,9,10,11,12,12a,13,13a-dechydro-3-methoxy-12-ethylsulfonyl-6h-isoquino 2,1-g 1,6 naphthyridine hydrochloride,4ar,12bs,13as-1-ethanesulfonyl-10-methoxy-2,3,4,4a,5,7,8,12b,13,13a-decahydro-1,6-diazatetraphene hydrochloride,8ar,12as,13as-12-ethylsulfonyl-5,8,8a,9,10,11,12,12a,13,13a-decahydro-3-methoxy-6h-isoquino 2,1-g 1,6 naphthyridine hydrochloride,8ar,12as,13as-5,8,8a,9,10,11,12,12a,13,13a-decahydro-3-methoxy-12-ethylsulfonyl-6h-isoquino 2,1-g 1,6 naphthyridine hydrochloride
9908991
(8aR,12aS,13aS)-12-ethylsulfonyl-3-methoxy-5,6,8,8a,9,10,11,12a,13,13a-decahydroisoquinolino[2,1-g][1,6]naphthyridine;hydrochloride
CCS(=O)(=O)N1CCCC2C1CC3C4=C(CCN3C2)C=C(C=C4)OC.Cl

Potent, selective α2 antagonist

α2-adrenoreceptor antagonist (Kd values are; 0.18, 0.19 and 0.42 nM for α2B α2C and α2A, respectively in rat).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 400.96
Percent Purity >99%
Chemical Name or Material RS 79948 hydrochloride

Recommended Storage : Store at Room Temperature