Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(RS)-Atenolol, Tocris Bioscience™
Click to view available options
Quantity:
1g
Description
Cardioselective β-adrenergic blocker. Antihypertensive, antianginal and antiarrhythmic.
S-enantiomer also available.
Specifications
Quantity | 1g |
Formula Weight | 266.34 |
Percent Purity | >98% |
Chemical Name or Material | (RS)-Atenolol |
Safety and Handling
Recommended Storage : Store at Room Temperature