Learn More
(RS)-PPG
Potent and selective mGlu8 agonist
Manufacturer:  Tocris Bioscience 1220/5
 View more versions of this product
Catalog No. 12-205
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
(RS)-PPG is a potent and selective group III metabotropic glutamate receptor agonist, with approximately 25-fold preference for hmGlu8; anticonvulsant and neuroprotective in vivo.
Specifications
Specifications
Observed MW: 231.14
Group III mGluR Agonists
120667-15-4
C8H10NO5P
(RS)-4-Phosphonophenylglycine
5 mg
Safety and Handling
Safety and Handling
Recommended Storage : Store at Room Temperature