Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSAD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549567

Catalog No. PA549567

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

RSAD1 may be involved in porphyrin cofactor biosynthesis.
Specifications

Specifications

RSAD1
Polyclonal
PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Q9HA92, Q5SUV1
Radical S-adenosyl methionine domain-containing protein 1, mitochondrial, 1399-, Oxygen-independent coproporphyrinogen-III oxidase-like protein RSAD1, RSAD1
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
237926, 55316, 688146
Immunohistochemistry, Western Blot
Unconjugated
RSAD1
Liquid
RSAD1
synthetic peptide corresponding to 311-342aa of human RSAD1
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit