Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSBN1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548230

Catalog No. PA548230

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

RSBN1
Polyclonal
PBS with 0.09% sodium azide
Q5VWQ0
Round spermatid basic protein 1, RSBN1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54665
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RSBN1
Liquid
RSBN1
KLH conjugated synthetic peptide between 51-78 amino acids from the N-terminal region of human RSBN1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit