Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RSBN1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | RSBN1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP157652
|
Novus Biologicals
NBP157652 |
100 μL |
Each of 1 for $436.00
|
|
Description
RSBN1 Polyclonal specifically detects RSBN1 in Human samples. It is validated for Western Blot.Specifications
RSBN1 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
DKFZp781E21150, FLJ11220, ROSBIN, round spermatid basic protein 1, RP11-324J2.1 | |
RSBN1 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
Q5VWQ0 | |
54665 | |
Synthetic peptides corresponding to RSBN1(round spermatid basic protein 1) Antibody(against the N terminal of RSBN1. Peptide sequence EGKEKPHAGVSPRGVKRQRRSSSGGSQEKRGRPSQEPPLAPPHRRRRSRQ. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title