Learn More
RSF1 Mouse anti-Human, Clone: C.15100041, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259281
Catalog No. NBP259281
Add to cart
Description
Description
RSF1 Monoclonal antibody specifically detects RSF1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunofluorescence.
Specifications
Specifications
RSF1
C.15100041
Western Blot 1:1000, Immunoprecipitation 1 ug, Immunofluorescence 1:100 - 1:500
Ascites
RSF1
The exact immunogen is propietary information.
Ascites
RUO
Primary
51773
Western Blot, Immunoprecipitation, Immunofluorescence
Unconjugated
Ascites Fluid with 0.05% Sodium Azide
HBV pX associated protein-8, HBXAPp325 subunit of RSF chromatin-remodeling complex, hepatitis B virus x associated protein, Hepatitis B virus X-associated protein, p325, remodeling and spacing factor 1, Rsf-1, XAP8HBV pX-associated protein 8
Mouse
IgG1
100 ul
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Monoclonal
Human