Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RSG1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | RSG1 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
RSG1 Polyclonal specifically detects RSG1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
RSG1 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
C1orf89, chromosome 1 open reading frame 89, MGC10731, miro domain-containing protein C1orf89, Rem/Rab-Similar GTPase 1, REM2 and RAB-like small GTPase 1, RP4-733M16.4 | |
RSG1 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
79363 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:VHHETTGIQTTVVFWPAKLQASSRVVMFRFEFWDCGESALKKFDHMLLACMENTDAFLFLFSFTDRASFEDLPGQLARIAGEAPGVVRMVIGS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title